Order Entry
ContactUsLinkComponent
Int-H1-S6A F8 Control Peptide
Int-H1-S6A F8 Control Peptide
Catalog #: 89161-288
Supplier:  Enzo Life Sciences
CAS Number:  
Int-H1-S6A F8 Control Peptide 0.5 Mg
Int-H1-S6A F8 Control Peptide
Catalog #: 89161-288
Supplier:  Enzo Life Sciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    0.5 mg
  • Protein/peptide name:
    Int-H1-S6A F8 control Peptide
  • Purity:
    95%
  • Tested applications:
    Control Peptide
  • Cat. no.:
    89161-288
  • Supplier No.:
    P606-0500

Specifications

About this item

Negative control for Int-H1-S6A F8

H1 DNA-binding region of c-Myc containing Ser to Ala and Phe to Ala substitutions (underlined) to confer an increase in its ability to inhibit c-Myc. It inhibits the CM1 consensus sequence binding of c-Myc-92 (C-terminus of c-Myc containing the bHLH region) with IC50= 1 µM. It is a non-cell-permeable control for Int-H1-S6A,F8A.

Customers Who Bought This Also Bought