Order Entry
ContactUsLinkComponent
p53 Activator
p53 Activator
Catalog #: 89161-284
Supplier:  Enzo Life Sciences
CAS Number:  
P53 Activator 0.5 Mg
p53 Activator
Catalog #: 89161-284
Supplier:  Enzo Life Sciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    0.5 mg
  • Protein/peptide name:
    p53 activator
  • Purity:
    97%
  • Cat. no.:
    89161-284
  • Supplier No.:
    P603-0500

Specifications

About this item

p53 activator

A peptide corresponding to 'peptide 46', (aa 361-382), which corresponds to the single-stranded DNA-end binding site of p53. This sequence is an activator of wild-type and certain p53 mutants. Fusion Peptide 46, a 35-mer peptide, consists of peptide 46 linked to a sequence from the third helix of Drosophila Antennapedia homeodomain protein in order to confer cell permeability. The p53 Activator (Fusion Peptide 46) has been demonstrated to have numerous effects including: rescue of p53 growth inhibition (5 µg/ml) in a human osteosarcoma Saos-2 subline transfected with His273 p53 mutant; at 20 µM, it induced 56% apoptosis in Burkitt lymphoma cell line Ew36 carrying wt p53; and it induced apoptosis in mutant p53 cell lines BL41 (20%) and SW480 (25%). It has no effect in p53 null cell lines HL60 and Saos-21. The p53 activator peptide is a useful tool to study p53.

Customers Who Bought This Also Bought