Specifications
- Size:5 mg
- Protein/peptide name:Ac-REQD-pNA
- Purity:95%
- Cat. no.:89161-248
- Supplier No.:P442-0005
Specifications
About this item
Caspase substrate
Colorimetric caspase substrate. Sequence corresponds to the prodomain of procaspase-8 cleavage site, residues 213-216, a putative caspase-10 target. Selective caspase-1 substrate at concentrations of 0.2-2 mM.