Specifications
- Size:1 mg
- Protein/peptide name:Z-Val-Ala-Asp(OMe)-fluoromethyl ketone
- Purity:>95%
- Cat. no.:89161-240
- Supplier No.:P416-0001
Specifications
About this item
Caspase inhibitor
Irreversible and cell permeable broad-spectrum caspase inhibitor. Inhibits Fas-mediated apoptosis in Jurkat T cells and apoptosis in THP.1 cells induced by diverse stimuli (including cycloheximide, thapsigargin, etoposide and staurosporine). For cell free assays use Prod. No. ALX-260-020.