Specifications
- Size:1 mg
- Protein/peptide name:Pp60V-Src Autophosphorylation Site
- Purity:97%
- Cat. no.:89161-208
- Supplier No.:P307-0001
Specifications
About this item
EGFR substrate
Residues 2-12 are the v-Src (412-422) autophosphorylation site with an extra Arg for phosphocellulose binding. This substrate sequence, RRLIEDNEYTARG, was originally described as an EGFR substrate in A431 cell extracts (Km=0.3-0.6 mM).