To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:5 mg
- Peptide content:60-80%
- Protein/peptide name:Boc-Ala-Ala-Nva-Sbzl (Chromogenic Substrate)
- Purity:>95% (HPLC)
- Cat. no.:89161-200
- Supplier No.:P303-0005
Specifications
About this item
Proteinase substrate
Much more sensitive substrate for leukocyte proteinase 3 (PR-3, myeloblastin) (Kcat/km=1.06 x 106 M-1s-1, Km=63 µM) than MeOSuc-AAPV-pNA (Chromogenic Substrate) (Prod. No. BML-P213) (Kcat/km=1.06 x 103 M-1s-1, Km=470 µM). Also cleaved by neutrophil elastase (Kcat/km=9.2 x 106 M-1s-1, Km=2.2 µM), granzyme H, cathepsin G, chymotrypsin, and rat MCPI and II. Typical assay conditions: substrate concentration 88-250 µM; enzyme concentration 0.2-20 nM; buffer [100 mM HEPES pH 7.5, 500 mM NaCl, 10% DMSO]; with 170 µM either 5,5'-dithiobis(2-nitrobenzoic acid) (DTNB) or 4,4'-dithiobis(pyridine) (from 15 mM DMSO stocks); 25°C. Hydrolysis of substrate can be detected at 412 nm for DTNB (