To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Biotin
- Size:100 µg
- Protein/peptide name:PDKtide
- Purity:95%
- Cat. no.:89161-158
- Supplier No.:P251-0100
Specifications
About this item
Protein kinase substrate
This peptide, Biotin-Ahx-KTFCGTPEYLAPEVRREPRILSEEEQEMFRDFDYIADWC, is derived from AKT1 and PKN2/PRK2. It is a substrate for PDK1, CDK9/Cyclin T1 and JAK2.The biotin allows peptide to be used in kinase assays with streptavidin-bound membranes.