Specifications
- Size:100 mg
- Protein/peptide name:Leu-Pna (Chromogenic Substrate)
- Purity:99%
- Cat. no.:89161-114
- Supplier No.:P230-0100
Specifications
About this item
Aminopeptidase substrate
Substrate for aminopeptidases such as aminopeptidase N (APN) cystinyl aminopeptidase, and leucine aminopeptidase (LAP). This substrate can be used for in vitro assays with purified enzyme, or with cell extracts. Also available: calibration standard pNA (BML-KI145).