To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:0.5 mg
- Protein/peptide name:PKC Inhibitors
- Purity:>97% (HPLC)
- Formulation:Lyophilized
- Cat. no.:89161-100
- Supplier No.:P219-0500
Specifications
About this item
PKC inhibitor.
Pseudosubstrate sequence from human PKCzeta (113-125) completely conserved in rabbit, rat, mouse and Xenopus. Sequence is also identical in human PKCι; (114-126) and mouse PKClambda. The N-terminus is myristoylated to allow cell permeability. Peptide inhibitor of atypical PKC's (calcium/phorbol ester/diacyglycerol insensitive). Useful in conjunction with the Myristoylated PKC (20-28), Cell-permeable PKC Inhibitor (BML-P205).