Specifications
- Size:0.5 mg
- Protein/peptide name:Protein Kinase Inhibitor
- Purity:>95% (HPLC)
- Cat. no.:89161-070
- Supplier No.:P203-0500
Specifications
About this item
PKA inhibitor
Sequence is from the heat-stable skeletal muscle inhibitor protein of PKA. Peptide binds to the catalytic subunit of PKA and displaces the regulatory subunit. Mimics protein substrate by binding to the catalytic site via the Arg-cluster basic residues. Ki =2.3 nM for PKA.