To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:10 mg
- Protein/peptide name:Z-VVR-AMC (Fluorogenic Substrate)
- Purity:>99% (TLC)
- Cat. no.:89161-062
- Supplier No.:P199-0010
Specifications
About this item
Substrate for cathepsin S and cathepsin L
Sensitive fluorogenic substrate for cathepsin S and cathepsin L. Also cleaved by cathepsin B, cathepsin V, kallikrein 8, and factor VIIa/TF but not cathepsin K nor kallikrein 2. Ex.: 380 nm, Em.: 460 nm, although the following Ex/Em can also be used: 355,375/440,450. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144).