To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:96 Wells
- Storage conditions:–80 °C (Long Term)
- Application:Colorimetric Detection, HTS Activity Assay
- Cat. no.:89156-550
- Supplier No.:AK804-0001
Specifications
About this item
This colorimetric kit is designed for inhibitor screening or kinetic assay of recombinant Calcineurin
- Safe and non-radioactive assay
- Convenient 1-step detection, no mixing needed
Activity is determined using the RII phosphopeptide substrate, the most efficient and selective peptide known for calcineurin, and detection of free phosphate released is based on the classic malachite green assay.
phosphate standard, microplate and detailed instructions.