To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:96 Wells
- Storage conditions:–80 °C (Long Term)
- Application:Colorimetric Detection, Fluorescent Detection, HTS Activity Assay
- Cat. no.:89156-540
- Supplier No.:AK703-0001
Specifications
About this item
This kit is a complete assay system for measuring caspase-3 and caspase-3-like activity (DEVDase) in cellular extracts.
It is performed in a 96-well format with all reagents for preparing cell extracts, measuring caspase activity and calibrating the assay. The caspase-3, is useful as a positive control, for testing cellular extracts for endogenous inhibitors, and more