To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Sensitivity:Can detect DPPIV activity in most or all biological samples
- Size:96 Wells
- Storage conditions:–80 °C (Long Term)
- Application:Colorimetric Detection, Fluorescent Detection, HTS Activity Assay
- Cat. no.:89156-498
- Supplier No.:AK498-0001
Specifications
About this item
The DPPIV/CD26 Assay kit for biological samples is a complete assay system designed to measure DPPIV activity in biological fluids such as plasma, serum, urine, and saliva. Uses for the kit include correlation of DPPIV activity with disease states or determination of the efficacy of DPPIV inhibitors administered in vivo.
Because DPPIV/CD26 protein levels may not accurately reflect the levels of active DPPIV enzyme, it is useful to measure DPPIV activity rather than performing DPPIV immunoassay. The kit can easily be used for other biological fluids such as tissue, live cells and cell extracts, and exudates
The fluorimetric assay is based on the cleavage of 7-amino-4-methylcoumarin (AMC) moiety from the C-terminus of the peptide substrate, which increases its fluorescence intensity at 460 nm. A specific DPPIV inhibitor, P32/98, is included to uncover any activity not contributed by DPPIV. Also included is DPPIV enzyme to use as a positive control.
Enough reagents are provided to perform at least 96 assays. The kit contains both a chromogenic substrate (H-Gly-Pro-pNA) and a fluorogenic substrate (H-Gly-Pro-AMC). Cleavage of the p-nitroaniline (pNA) from the chromogenic substrate increases absorbance at 405 nm.