To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:1 Kit
- Storage conditions:−80 °C
- Application:Colorimetric Detection, HTS, Activity Assay
- Cat. no.:89156-458
- Supplier No.:AK404-0001
Specifications
About this item
A QuantiZymeTM Assay System
The MMP-1 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-1 (MMP-1, interstitial collagenase, fibroblast collagenase) using a thiopeptide as a chromogenic substrate.