Specifications
- Size:1 Kit
- Cat. no.:89156-418
- Supplier No.:AK016-0001
- No. of Tests:96 Tests
Specifications
About this item
A QUANTIZYME® Assay System Complete assay system designed to examine the specificity of inhibitors against a panel of ten matrix metalloproteinase enzymes. The assays are performed in a convenient 96-well microplate format using a sensitive fluorogenic substrate . Included are active MMP enzymes, assay buffer, a prototypic control inhibitor (NNGH3), and a detailed instruction booklet.