To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:100 Tests
- Storage conditions:–80 °C (Long Term)
- Application:Colorimetric Detection, Activity Assay
- Cat. no.:89156-146
- Supplier No.:850-326-KI01
Specifications
About this item
Histone acetyltransferases (HATs) have been implicated to play a crucial role in various cellular functions, such as gene transcription, differentiation, and proliferation
The kit utilizes active recombinant HAT as a positive control and acetyl-CoA as a cofactor. Acetylation of peptide substrate by active HAT releases the free form of CoA which then serves as an essential coenzyme for producing NADH. NADH can easily be detected spectrophotometrically upon reacting with a soluble tetrazolium dye. The detection can be continuous and suitable for kinetic studies. The kit provides a simple, straightforward protocol for a complete assay.
HAT Activity Colorimetric Assay Kit offers a convenient, nonradioactive system for a rapid and sensitive detection of HAT activity in mammalian samples.