Specifications
- Size:1 Kit
- Storage conditions:–20 °C (Long Term)
- Cat. no.:89156-084
- Supplier No.:850-277-KI01
Specifications
About this item
For the detection of Cytochrome c translocation from mitochondria into cytosol during apoptosis.
The kit provides an easy-to-use procedure and reagents for separating a mitochondria-enriched fraction from cytosol. Cytochrome c released from mitochondria into cytosol is determined by Western blot using the cytochrome c antibody provided in the kit. The procedure is simple, straightforward and requires no ultracentrifugation.