To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:100 Tests
- Storage conditions:–20 °C (Long Term)
- Species reactivity:Mammalian
- Cat. no.:89156-082
- Supplier No.:850-276-KI01
Specifications
About this item
For the effective isolation of a highly enriched mitochondrial fraction from the cytosolic fraction of various mammalian cells including both apoptotic an non-apoptotic cells
The core kit contains unique formulations of buffers and reagents that allow isolation of mitochondria and cytosol in only two easy steps. No ultracentrifugations are required. No toxic chemicals are involved.
The enriched mitochondrial and cytosolic fractions can be used to study apoptosis and signal trasduction pathways by detecting translocations of factors of interest between the two fractions by ELISA, Western blot or other assays.