Order Entry
ContactUsLinkComponent
Caspase-8 Colorimetric Assay Kit, Enzo Life Sciences
Caspase-8 Colorimetric Assay Kit, Enzo Life Sciences
Catalog #: 89155-970
Supplier:  Enzo Life Sciences
CAS Number:  
Caspase-8 Colorimetric Assay Kit 25 Tests
Caspase-8 Colorimetric Assay Kit, Enzo Life Sciences
Catalog #: 89155-970
Supplier:  Enzo Life Sciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    25 tests
  • Storage conditions:
    –20 °C (Long Term)
  • Application:
    Colorimetric Detection, Activity Assay
  • Cat. no.:
    89155-970
  • Supplier No.:
    850-221-KI01

Specifications

About this item

Based on the spectrophotometric detection of the chromophore pNA (p-nitroaniline) after cleavage from the labelled substrate IETD-pNA.

Ordering information: Contents: 25ml Cell Lysis Buffer, 2ml 2X Reaction Buffer, 125µl IETD-pNA Substrate (4mM), 100µl DTT (1M), 25ml Dilution Buffer.
Delivery: Kit includes Cell lysis buffer (25 mL), 2X Reaction buffer(2 mL), 4 mM IETD-pNA Substrate (125 µL), 1 M DTT (100 µL), Dilution bBuffer (25 mL).
Customers Who Bought This Also Bought