Annexin V-FITC Apoptosis Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
Certificates
About this item
For the detection of apoptotic cells by flow cytometry.
FITC-labelled recombinant human annexin V shows bright green fluorescence (Ex(max): 488 nm, Em(max): 530 nm).
Delivery: Kit contains Annexin V-FITC, binding buffer and propidium iodide.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.