To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD14 Molecule
- Clonality:Monoclonal
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:CD14
- ELISA:Yes
- Flow cytometry:Yes
- Size:100 µg
- Western blot:Yes
- Form:Lyophilized from PBS. Contains no preservatives.
- Antigen synonyms:monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein
- Concentration:1mg/ml after reconstitution.
- Purification:Protein G affinity-purified
- Cat. no.:89154-396
- Supplier No.:804-496-C100
Specifications
About this item
The CD14 glycoprotein, gp 55, is present on most monocytic and macrophage like cell types: monocytes, macrophages, Kupffer cells, pleural phagocytic cells and dendritic reticular cells. CD14 is also observed on granulocytes and activated or transformed B-cells. Furthermore CD14 is present in a soluble form in human serum, urine and other body fluids. The CD14 molecule has been reported to be a receptor for endotoxin.
- For ELISA, Flow Cytometry, Western Blot
Recognizes human (epitope aa 9-13 & aa 39-44), pig, dog, horse and bovine CD14.
Reconstitute with 100µl distilled water.
Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human