To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:NA+/K+ ATPase alpha
- Clonality:Monoclonal
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:400 µL
- Western blot:Yes
- Form:Liquid. In 400µl PBS containing 1mg/ml BSA and 0.05% sodium azide.
- Purification:Protein A-affinity purified
- Cat. no.:89153-632
- Supplier No.:804-081-1
Specifications
About this item
Host: Mouse, Isotype: IgG1
- For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes human, mouse, rat (alpha3 isoform only), sheep, bovine, pig, dog and chicken Na+/K+ ATPase-alpha. Detects a band of ~110kDa by Western blot.
Type: Primary
Antigen: [delete]
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human