To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Microtubule-associated Protein 1 light chain 3 beta
- Clonality:Monoclonal
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:LC3B
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Form:Liquid. In PBS containing 50% glycerol, 0.09% sodium azide, PEG and sucrose.
- Antigen synonyms:MAP1LC3B|Autophagy-related protein LC3 B|MAP1 light chain 3-like protein 2|MAP1ALC3|MAP1A/MAP1B LC3 B|Microtubule-associated proteins 1A/1B light chain 3B [Precursor]|MAP1A/1B light chain 3 B|Autophagy-related ubiquitin-like modifier LC3 B
- Concentration:0.1mg/ml
- Purification:Thiophilic adsorption and size exclusion chromatography purified
- Cat. no.:89153-436
- Supplier No.:803-081-C100
Specifications
About this item
Autophagy is an alternative process of proteasomal degradation for some long-lived proteins or organelles. Alterations in the autophagic-lysosomal compartment have been linked to neuronal death in many neurodegenerative disorders as well as in transmissible neuronal pathologies (prion diseases). Genetic studies in yeast have shown that Autophagy-defective Gene-8 (Atg-8) represents a specific marker for autophagy. Among the four families of mammalian Atg8-related proteins only LC3 (Microtubule-associated Protein1 Light Chain 3) is expressed at sufficient high levels and efficiently recruited to autophagic vesicles in cells and tissues. During autophagy the cytoplasmic form, LC3-I is processed and recruited to autophagosomes, where LC3-II is generated by site specific proteolysis near to the C-terminus. Autophagic vacuoles have been also reported frequently in cardiomyopathies or muscle cells exposed to different experimental settings.
- For Immunocytochemistry, Western Blot
Recognizes human, mouse, rat, monkey and hamster LC3B. Recognizes both forms of endogenous LC3B. Detects a band of ~18kDa (LC3B-I; cytoplasmic form) and a band of ~16kDa (LC3B-II; lipidated form) by Western blot.
Type: Primary
Antigen: LC3B
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human