To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:alpha Gal Epitope
- Clonality:Monoclonal
- Host:Mouse
- Isotype:IgM
- Reactivity:Mouse
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Size:1 ViaL
- Western blot:Yes
- Form:Liquid. Tissue culture supernatant.
- Cat. no.:89153-336
- Supplier No.:801-090-1
Specifications
About this item
Host: Mouse, Isotype: IgM
- For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Recognizes synthetic and naturally produced mouse, rat and pig Galalpha1-3Gal epitopes on glycoproteins and glycolipids. Does not cross-react with beta-Gal glycoproteins or BSA.
Type: Primary
Antigen: [delete]
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Mouse