Order Entry
ContactUsLinkComponent
Mouse Recombinant MegaCD40L™ (soluble) (from CHO cells)
Mouse Recombinant MegaCD40L™ (soluble) (from CHO cells)
Catalog #: 89151-916
Supplier:  Enzo Life Sciences
CAS Number:  
MegaCD40L* SOLUBLE (Recombinant.) 10 uG
Mouse Recombinant MegaCD40L™ (soluble) (from CHO cells)
Catalog #: 89151-916
Supplier:  Enzo Life Sciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Mouse
  • Size:
    10 µg
  • Endotoxin content:
    \<0.1EU/µg purified protein (LAL test; Bio Whittaker).
  • Biological activity:
    Binds to mouse and human CD40. Induces B cells activation (as demonstrated by dose-dependent upregulation of CD86).
  • Reconstitution instructions:
    Reconstitute with 100µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum.
  • Protein/peptide name:
    MegaCD40L™ (soluble)
  • Purity:
    >95% (SDS-PAGE)
  • Concentration:
    0.1mg/ml after reconstitution.
  • Formulation:
    Lyophilized. Contains PBS.
  • Cat. no.:
    89151-916
  • Supplier No.:
    522-120-C010

Specifications

About this item

Highly active. Produced in CHO cells. The extracellular domain of mouse CD40L (CD154) (aa 115-260) is fused at the N-terminus to mouse ACRP30headless (aa 18-111) and a FLAG®-tag.

MegaCD40L™ is a high activity construct in which two trimeric CD40 ligands are artificially linked via the collagen domain of ACRP30/adiponectin. This construct very effectively simulates the natural membrane-assisted aggregation of CD40L. It provides a simple and equally potent alternative to [CD40L+enhancer] combinations (Prod. No. ALX-850-075).

Customers Who Bought This Also Bought