Z-WEHD-FMK (Ready-to-Use), Enzo Life Sciences
Supplier: Enzo Life Sciences
Certificates
About this item
Inhibitor of caspase-1, -4 and -5
Irreversible, cell permeable inhibitor of caspase-1 and -5. Inhibits also caspase-4. Designed as a methyl ester to facilitate cell permeability.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.