Z-VAD-FMK (Ready-to-Use), Enzo Life Sciences
Supplier: Enzo Life Sciences
Certificates
About this item
Caspase inhibitor
Synthetic peptide that irreversibly inhibits ICE-family protease/caspase activity and blocks apoptosis. The inhibitor is designed as a methyl ester to facilitate cell permeability.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.