About this item
Potent and cell permeable proteasome inhibitor. Inhibits the chymotrypsin-like activity of the 26S proteasome (MCP; multicatalytic proteinase complex). Does not inhibit the peptidylglutamyl-peptide hydrolyzing activity of the MPC.
White to off-white powder.
Soluble in DMSO.