To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:8-Hydroxyguanosine
- Clonality:Polyclonal
- Host:Goat
- Reactivity:Multiple
- Antigen symbol:8-Hydroxyguanosine
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 µL
- Form:Liquid. Neat serum.
- Cat. no.:89146-090
- Supplier No.:210-802-R100
Specifications
About this item
8-Hydroxydeoxyguanosine (8OHdG) is a modified base that occurs in DNA due to attack by hydroxyl radicals that are formed as byproducts and intermediates of aerobic metabolism and during oxidative stress. There is increasing evidence to support the involvement of free radical reactions in the damage of biomolecules that eventually lead to several diseases in humans, such as atherosclerosis, cerebral and heart ischemia-reperfusion injury, cancer, rheumatoid arthritis, inflammation, diabetes, aging and neurodegenerative conditions, such as Alzheimer's disease. 8OHdG has become increasing popular as a sensitive, stable and integral marker of oxidative damage in cellular DNA. Biomonitoring in humans has demonstrated that 8OHdG can be excreted in the urine and that a significant increase is caused by exposure to tobacco smoke and ionizing radiation. Because 8OHdG is so well correlated with oxidative stress and damage to DNA, which leads to degenerative disease states, the development of an antibody that can be used to study DNA damage has numerous applications. In addition to the direct study of DNA damage within cells, this antibody has applications in the development of immunoassays that can monitor 8OHdG excretion in the urine and serve as a biomarker of oxidative stress. Industrial uses may extend to the dietary supplement manufacturers, who could benefit from an immunoassay that could be used to test the effectiveness of antioxidants and other nutraceuticals.
- For ELISA, Flow Cytometry, Immunohistochemistry
Recognizes 8-hydroxyguanosine and 8-hydroxy-2'-deoxyguanosine. Partially cross-reacts with 8-mercaptoguanosine and slightly with 8-bromoguanosine. Does not cross-react with other naturally occuring nucleotides.
Type: Primary
Antigen: 8-HYDROXYGUANOSINE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Species Independent