Human Recombinant Hepsin
Catalog #: 89144-974
Supplier: Enzo Life Sciences
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Size:125 U
- Biological activity:Trypsin-like serine protease.
~2.5U/µl. One unit is defined at the amount of enzyme that releases 1nmol of AMC per min. at 37°C in the presence of 50µM Boc-Gln-Ala-Arg-AMC. - Protein/peptide name:Hepsin
- Concentration:100nM
- Formulation:Liquid. In 100mM TRIS, pH 9.0, containing 200mM sodium chloride and 10% glycerol.
- Cat. no.:89144-974
- Supplier No.:201-750-1
Specifications
About this item
Produced in Drosophila Schneider 2 (S2) cells. Recombinant human hepsin (aa 45-417) is fused at the C-terminus to a V5-His-tag.
Customers Who Bought This Also Bought