Human Recombinant Leptin (from E. coli)
Catalog #: 89143-978
Supplier: Enzo Life Sciences
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:1 mg
- Endotoxin content:\<0.1EU/µg purified protein.
- Biological activity:Induces proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor.
- Reconstitution instructions:Reconstitute with sterile water or 0.4% sodium bicarbonate, pH 8-9. Do not reconstitute to less than 0.1mg/ml. Further dilutions should be made with medium containing 0.1% HSA or BSA.
- Protein/peptide name:Leptin
- Purity:>98% (SDS-PAGE)
- Formulation:Lyophilized from 0.0045mM sodium bicarbonate.
- Cat. no.:89143-978
- Supplier No.:201-034-M001
Specifications
About this item
Produced in E. coli. Homologous to human serum leptin.
Ordering information: Manufactured by BioVendor.
Customers Who Bought This Also Bought