To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Protein/peptide type:Native
- Source:Brain
- Species:Bovine
- Size:1 mg
- Protein/peptide name:MARCKS protein (159-165) amide
- Purity:>98%
- Cat. no.:89143-534
- Supplier No.:162-005-M001
Specifications
About this item
PKC substrate
The peptide consists of residues 159-165 of the myristoylated alanine-rich C-kinase substrate (MARCKS) protein. It is sufficiently selective for protein kinase C (PKC) to permit specific assay of this enzyme in crude cell extracts and permeabilized cells.