NG,NG'-Dimethyl-L-arginine dihydrochloride ≥98% (by TLC)
Catalog #: 89142-856
Supplier: Enzo Life Sciences
CAS Number: 30344-00-4
Specifications
- Size:25 mg
- Cat. no.:89142-856
- Supplier No.:106-008-M025
Specifications
Specification Test Results
- Purity:≥98% (TLC)
- NG,NG'-Dimethyl-L-arginine dihydrochloride ≥98% (by TLC):
Specification Test Results
About this item
Synonyms: SDMA dihydrochloride,SDMA.2HCl,Symmetric dimethylarginine dihydrochloride
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 275.2 g/mol
Formula:C₈H₁₈N₄O₂·2HCl
MW:275.2 g/mol
Storage Temperature:Freezer
CAS Number:30344-00-4
Customers Who Bought This Also Bought