NG, NG-Dimethyl-L-arginine dihydrochloride ≥97% (by TLC)
Supplier: Enzo Life Sciences
Certificates
About this item
Synonyms: H-Arg(di-Me)-OH.2HCl (asymmetrical)
Formula:C₈H₁₈N₄O₂·2HCl·H₂O
CAS Number:220805-22-1
:Length:
:Width:
:Height:
:Ø
:Thickness:
:97
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 293,2 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Purity:≥97% (TLC)
- NG, NG-Dimethyl-L-arginine dihydrochloride ≥97% (by TLC):
Specification Test Results
Customers Who Bought This Also Bought