S-Trityl-L-cysteine
Catalog #: 89142-820
Supplier: Enzo Life Sciences
CAS Number: 2799-07-7
Specifications
- Size:500 mg
- Cat. no.:89142-820
- Supplier No.:105-011-M500
Specifications
Specification Test Results
- S-Trityl-L-cysteine:
Specification Test Results
About this item
Synonyms: H-Cys(Trt)-OH
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 363.5 g/mol
Formula:C₂₂H₂₁NO₂S
MW:363.5 g/mol
MDL Number:MFCD00002611
CAS Number:2799-07-7
Customers Who Bought This Also Bought