N-Methyl-D-aspartic acid ≥98% (by TLC)
Catalog #: 89142-812
Supplier: Enzo Life Sciences
CAS Number: 6384-92-5
Specifications
- Size:50 mg
- Cat. no.:89142-812
- Supplier No.:105-009-M050
Specifications
Specification Test Results
- Purity:≥98% (TLC)
- Identity:Determined by NMR
- N-Methyl-D-aspartic acid ≥98% (by TLC):
Specification Test Results
About this item
Synonyms: N-Methyl-D-asparaginic Acid,Me-D-Asp-OH
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 147.13 g/mol
:N-Methyl-D-aspartic acid
Storage Temperature:Ambient
:14,06662
Formula:C₅H₉NO₄
MW:147.13 g/mol
MDL Number:MFCD00004226
CAS Number:6384-92-5
Customers Who Bought This Also Bought