To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Discs Large MAGUK Scaffold Protein 4
- Clonality:Monoclonal
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Mouse
- Antigen symbol:DLG4
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Antigen synonyms:PSD-95 |SAP90|FLJ98574|PSD95|SAP-90 |FLJ97752
- Cat. no.:CA200062-880
- Supplier No.:VAM-PS002E
Specifications
About this item
Postsynaptic density protein 95 kDa (PSD-95), also known as Synapse associated protein 90 kDa (SAP90), is a brain specific protein that is highly similar to the Drosophila dlg tumor suppressor protein. PSD-95 is a member of membrane associated proteins that are localized beneath the postsynaptic membrane of synapses in the CNS. PSD-95 interacts with NMDA receptor and Shaker-type K+ channels and contributes to their clustering and localization at the synaptic spines in hippocampal neurons and the pinceau terminal of cerebellar basket cells, respectively.
- For Immunocytochemistry, Western Blot
Recognizes mouse, rat and bovine PSD-95. Detects a band of ~100kDa by Western blot.
Type: Primary
Antigen: DLG4
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse