Fixation/Permeabilization, (buffer), Enzo Life Sciences
Catalog #: 89141-772
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:100 mL
- Purity detail:Purified
- Formulation:Liquid. In PBS, pH 7.2, containing 4% formaldehyde and 0.1% Saponin.
- Cat. no.:89141-772
- Supplier No.:950-010-100
Specifications
About this item
Customers Who Bought This Also Bought