To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Sensitivity:0.00195 units/mL
- Description:Myeloperoxidase fluorometric detection kit
- Size:500 tests
- Storage conditions:+4 °C (long term)
- Species reactivity:Species independent
- Cat. no.:89141-684
- Application:Fluorescence microscopy
- Supplier No.:907-029
Specifications
About this item
The Myeloperoxidase fluorometric detection kit is a one-step no wash homogeneous activity assay that is adaptable to HTS applications
- Fast homogenous assay
- Suitable for High Throughput Screening
- Monitor multiple time points to follow kinetics
- Results in 30 to 60 minutes
For the measurement of Myeloperoxidase activity in cells and tissue from any species.
This kit offers highly sensitive fluorescent detection and the ability to monitor multiple time points to follow the kinetics of MPO activity.