To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:30 ml
- Storage temperature:4 °C (Long term)
- Cat. no.:CA95045-198
- Supplier No.:80-1929
Specifications
About this item
This Tween®-20 solution (10%) is specially designed to create the Blocking Buffer and Assay Buffer in ImmunoSet products. Each bottle is sufficient for use with 5×96 wells microtiter plates if following the instructions in our ImmunoSet protocol.
The Tween-20 solution may be used for any other assay development needs. Additional assay development reagents are available as companion products to our ImmunoSet products.