About this item
The Thermo Scientific Megafuge™ 8 centrifuge provides exceptional capacity in a compact design with a smart, simple interface and the flexibility to support both clinical and research applications. The centrifuge fits in to space, with its compact footprint, and to applications, accomodating multi-laboratory settings.
The Heraeus® Megafuge® 8 centrifuge stands out with innovative new technologies. Auto-Lock™ rotor exchange enables secure, push-button application versatility and cleaning convenience. ClickSeal™ biocontainment lids provide one-handed, certified sample protection. The unit offers maximized capacity, with the ability to process 24x5/7mL blood tubes, 8x50mL conical tubes, microplates and microtubes, all in one versatile unit. The Heraeus® Megafuge® 8 R has the fixed angle flexibility to spin 50mL conical tubes at over 12,000g, or a wide range of microtubes at speeds up to 30,279g.
The Megafuge™ 8 centrifuge stands out with innovative new technologies
The Megafuge™ 8 fits in to clinical applications, including clinical chemistry, clinical microbiology, hematology, immunology, and clinical studies. It runs up to 24 x 5/7mL blood or urine tubes at a time in swinging bucket configuration. The unit fits in to research applications as well, including cellular biology, microbiology, genomics/molecular biology, proteomics, biochemistry, and pharmaceutical studies. It runs up 8 x 50 mL swinging bucket rotor capacity, 4 microplates, or 30 filtration columns.
Additional features on this centrifuge include a quiet performance (<61 dBA ventilated and <52 dBA refrigerated), one-touch operation with pre-saved protocols and password protection, and a highly visible backlit display. All units are glove- and detergent-friendly.
Auto-Lock™ rotor exchange enables secure, push-button application versatility and cleaning convenience. ClickSeal™ biocontainment lids provide one-handed, certified sample protection. The unit offers maximized capacity, with the ability to process 24x5/7mL blood tubes, 8x50mL conical tubes, microplates and microtubes, all in one versatile unit. The Megafuge™ 8 R has the fixed angle flexibility to spin 50mL conical tubes at over 12,000g, or a wide range of microtubes at speeds up to 30,279g.
Specifications
- Catalog No:
- CA10067-600
- 89511-734
- 89423-731
- 89423-733
- Acceleration/braking profiles:
- 2 (standard and soft)
- 2 (standard and soft)
- Control:
- Microprocessor
- Microprocessor
- Memory:
- 4 direct programs with alphanumeric program naming; Refrigerated unit: 3 direct program buttons plus 96 additional programs accessible via folder all with alphanumeric program naming
- 4 direct programs with alphanumeric program naming; Refrigerated unit: 3 direct program buttons plus 96 additional programs accessible via folder all with alphanumeric program naming
- Display:
- Digital
- Digital
- Temperature setting range:
- –10 to 40°C (14 to 104°F) [Refrigerated Unit]
- –10 to 40°C (14 to 104°F) [Refrigerated Unit]
- Depth:
- 48 cm (18.9"); 67 cm (26.4") [Refrigerated Unit]
- 48 cm (18.9"); 67 cm (26.4") [Refrigerated Unit]
- Width:
- 37 cm (14.6"); 46 cm (18.1") [Refrigerated Unit]
- 37 cm (14.6"); 46 cm (18.1") [Refrigerated Unit]