To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Large Display Conductivity Pen
- Dimensions:165 × 38 × 38 mm (6.5 × 1.5 × 1.5")
- Weight:68 g (2½ oz.)
- Cat. no.:97051-722
- Supplier No.:850037
Specifications
About this item
Highly stable and accurate readings with large LCD display
Designed for simple one-hand operation. Simultaneously displays the parameter being measured and temperature in °C or °F. Features automatic temperature compensation, 2-pt. calibration, auto power off, hold function, and a low battery indicator. The case is IP65 waterproof, and will float if accidentally dropped into the water.