Specifications
- Chemical method:Chlorophenol red
- Description:Chlorine Dioxide Reagent Set, Chlorophenol Red
- Method:8065 - Chlorophenol Red
- Cat. no.:CA11024-830
- Range:0.01 - 1.00 mg/l ClO₂
- Supplier No.:2242300
- No. of Tests:100
Specifications
About this item
For determination of Chlorine Dioxide by the Chlorophenol Red Method
Hach method 8065. Range: 0.01 - 1.00 mg/L ClO2. Set contains Chlorine Dioxide Reagent 1, 2, and 3, and Dechlorinating Reagent Powder Pillows for 100 tests. Sample Size: 50 mL.