To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- W×L:152×229 mm (6×9")
- Color:Red Stat
- Description:Biohazard specimen bag
- Cat. no.:56766-390
- Packaging:50/bundle
- Supplier No.:SBL2X69S
Specifications
About this item
Bold RED Color and STAT verbiage ensure that specimens packed inside are handled with the utmost care and processed quickly.
Bags are packed in easy-to-use DispenserBag® Convenience Packaging.
Bags are packed in easy-to-use DispenserBag® convenience packaging.