Specifications
- Description:Ward's® AP Chemistry Investigation 15: Buffering
- No. of Lab Groups:15
- Cat. no.:470303-024
- Includes:Teacher's Guide and Student Copymaster
- Supplier No.:470303-024
Specifications
About this item
Meets AP Science Practices 4, 5, and 6
- Addresses Big Ideas 1 & 6
- Materials provided for 30 students working in pairs
- Investigation includes updated literature aligned with new AP Chemistry curriculum standards
In this lab, students will observe the buffering abilities of a number of different household products. Students will use the concepts of acid-base chemistry, titrations, pH, and buffers in order to determine if certain household products will act as buffers, and if so which chemicals are present. Titration curves will be constructed for known chemicals and then compared to the titration curves produced for the household products. Includes sodium hydroxide, citric acid, diet coke, diet lemon-lime soda, lemonade, grape juice, phosphoric acid, and acetic acid. Requires pH meter, ring stand/clamps, magnetic stirrer, magnetic stir rod, and burette, available separately. Ward's® DataHub not required to complete activity.
AP and Advanced Placement Programs are registered trademarks of the College Entrance Examination Board, which was not involved in the production of and does not endorse these products.
Includes:
Sodium Hydroxide
Hydrochloric Acid
Citric Acid
Phosphoric Acid
Acetic Acid
Diet Coke
Diet Sprit
Lemonade
Grape Juice
Required but Not Provided: pH Meter, Beakers, Pipettes, Graduated Cylinders, Erlenmeyer Flasks, Ring Stands/Clamps, Burettes, DI Water, Gloves, Safety Goggles, Lab Aprons.