Specifications
- Description:Ward's® AP Chemistry Investigation 14: Acid-Base Titrations
- No. of Lab Groups:15
- Cat. no.:470303-022
- Includes:Teacher's Guide and Student Copymaster
- Supplier No.:470303-022
Specifications
About this item
Meets AP Science Practices, 1, 2, 3, 4, 5, 6, and 7
- Addresses Big Ideas 1 & 6
- Materials provided for 30 students working in pairs
- Investigation includes updated literature aligned with new AP Chemistry curriculum standards
In this lab, acid-base titrations will be performed in order to investigate the relationship between pH, concentration, and the structure of acids and bases. Students will carry out multiple acid-base titrations and will use a pH meter to monitor the pH of the resulting solutions. Students will then construct titration curves. Using known information and the data that was gathered, students will ultimately be able to determine the concentration of the acid or base, the ionization constant, and the percent ionization. Includes hydrochloric acid, acetic acid, sodium hydroxide, and phenolphthalein. Requires pH meter, burette, ring stand/clamps, magnetic stirrer, and magnetic stir bar, available separately. Ward's® DataHub is not required to complete activity.
AP and Advanced Placement Programs are registered trademarks of the College Entrance Examination Board, which was not involved in the production of and does not endorse these products.