Specifications
- Description:Ward's® AP Chemistry Investigation 9: Liquid-Liquid Extraction
- No. of Lab Groups:15
- Cat. no.:470303-042
- Includes:Teacher's Guide and Student Copymaster
- Supplier No.:470303-042
Specifications
About this item
Meets AP Science Practices 1, 4, and 6
- Addresses Big Idea 3
- Materials provided for 30 students working in pairs
- Investigation includes updated literature aligned with new AP Chemistry curriculum standards
Liquid-liquid extraction is a method used for the separation of a mixture using two immiscible solvents. The ability to separate the different compounds in a mixture using this technique depends upon how differently the compounds of the sample mixture partition themselves between the two solvents. In this laboratory activity, students will explore liquid-liquid extraction techniques and ultimately separate and identify the components of an unknown pill powder. Includes salicylic acid and acetanilide powder, ethyl acetate, hydrochloric acid, anhydrous magnesium sulfate, sodium bicarbonate solution, and sodium chloride solution. Requires hotplate, separately funnel, Buchner funnel, glass funnel, side arm flask, vacuum filter or aspirator, filter paper, analytical balance, Excedrin, watch glass and drying oven (optional), available separately.
AP and Advanced Placement Programs are registered trademarks of the College Entrance Examination Board, which was not involved in the production of and does not endorse these products.