3-Maleimidopropionic acid
Supplier: Bachem Americas
Certificates
About this item
Synonyms: 3-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoic acid,3-Maleimido-propionic acid,N-(2-Carboxyethyl)maleimide,MPS-Acid
Formula:C₇H₇NO₄
MW:169.14 g/mol
MDL Number:MFCD00043030
CAS Number:7423-55-4
:3-Maleimidopropionic acid
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 169.14 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought