NF31
Catalog #: Q-2610.0050BA
Supplier: Bachem Americas
CAS Number: 253426-51-6
Specifications
- Size:50 mg
- Cat. no.:Q-2610.0050BA
- Supplier No.:Q-2610.0050
Specifications
About this item
Synonyms: (2-{Ethyl-[4-(4-nitro-phenylazo)-phenyl]-amino}-ethoxy)-acetic acid-4-nitro-phenyl ester
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 493.476 g/mol
:NF31
Formula:C₂₄H₂₃N₅O₇
MW:493.476 g/mol
Storage Temperature:Refrigerator
MDL Number:MFCD03093401
CAS Number:253426-51-6
Customers Who Bought This Also Bought